missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pentafluorobenzoate, 98%

Catalog No. AAL1052314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1052314 25 g
AAL1052306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1052314 Supplier Thermo Scientific Chemicals Supplier No. L1052314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4522-93-4 | C9H5F5O2 | 240.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4522-93-4
Molecular Formula C9H5F5O2
Molecular Weight (g/mol) 240.13
MDL Number MFCD00039211
InChI Key DFUDMSIRGGTHGI-UHFFFAOYSA-N
Synonym ethyl pentafluorobenzoate, ethyl perfluorobenzoate, pentafluorobenzoic acid ethyl ester, 2,3,4,5,6-pentafluoro-benzoic acid ethyl ester, benzoic acid, pentafluoro-, ethyl ester, ethylpentafluorobenzoate, acmc-1ajj1, rarechem al bi 0017, perfluorobenzoic acid ethyl ester
PubChem CID 78283
IUPAC Name ethyl 2,3,4,5,6-pentafluorobenzoate
SMILES CCOC(=O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Density 1.431
Boiling Point 112°C to 114°C
Flash Point 80°C (176°F)
Refractive Index 1.428
Quantity 25 g
Beilstein 2054770
Formula Weight 240.13
Percent Purity 98%
Chemical Name or Material Ethyl pentafluorobenzoate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 224-854-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only