missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Pentadecanoate 97.0+%, TCI America™

Catalog No. p-7134820
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08685ML 5 mL
P086825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08685ML Supplier TCI America Supplier No. P08685ML
Only null left

Chemical Identifiers

CAS 41114-00-5
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.46
MDL Number MFCD00042872
InChI Key PTEYJUIKYIKULL-UHFFFAOYSA-N
Synonym pentadecanoic acid, ethyl ester, n-pentadecanoic acid ethyl ester, unii-hj2a096y4t, pentadecanoic acid ethyl ester, ethyl=pentadecanoate, acmc-209jim, pentadecanoic acid,ethyl ester, ethyl pentadecanoate
PubChem CID 38762
IUPAC Name ethyl pentadecanoate
SMILES CCCCCCCCCCCCCCC(=O)OCC

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 mL
Formula Weight 270.46
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Pentadecanoate
EINECSNumber (2)-0798
TSCA No