accessibility menu, dialog, popup

UserName

CAS RN 41114-00-5

H3COOCH3

CAS RN 41114-00-5

IUPAC Name: ethyl pentadecanoate
Synonyms: ethyl pentadecanoate
Molecular Weight (g/mol): 270.46
Molecular Formula: C17H34O2
InChi Key: PTEYJUIKYIKULL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results