missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Palmitate 97.0+%, TCI America™
$46.60 - $301.98
Chemical Identifiers
| CAS | 628-97-7 |
|---|---|
| Molecular Formula | C18H36O2 |
| Molecular Weight (g/mol) | 284.48 |
| MDL Number | MFCD00008996 |
| InChI Key | XIRNKXNNONJFQO-UHFFFAOYSA-N |
| Synonym | ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm |
| PubChem CID | 12366 |
| ChEBI | CHEBI:84932 |
| IUPAC Name | ethyl hexadecanoate |
| SMILES | CCCCCCCCCCCCCCCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P000325ML
|
TCI America
P000325ML |
25 mL |
Each for $46.60
|
|
|||||
|
P0003500ML
|
TCI America
P0003500ML |
500 mL |
Each for $301.98
|
|
|||||
Chemical Identifiers
| 628-97-7 | |
| 284.48 | |
| XIRNKXNNONJFQO-UHFFFAOYSA-N | |
| 12366 | |
| ethyl hexadecanoate |
| C18H36O2 | |
| MFCD00008996 | |
| ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm | |
| CHEBI:84932 | |
| CCCCCCCCCCCCCCCC(=O)OCC |
Specifications
| 628-97-7 | |
| C18H36O2 | |
| 25 mL | |
| XIRNKXNNONJFQO-UHFFFAOYSA-N | |
| ethyl hexadecanoate | |
| 12366 | |
| 284.48 | |
| Crystalline Lumps |
| White-Yellow | |
| MFCD00008996 | |
| ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm | |
| CCCCCCCCCCCCCCCC(=O)OCC | |
| 284.48 | |
| CHEBI:84932 | |
| ≥97.0% (GC) | |
| Ethyl Palmitate |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes