accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 628-97-7

IUPAC Name: ethyl hexadecanoate
Synonyms: ethyl palmitate
Molecular Weight (g/mol): 284.48
Molecular Formula: C18H36O2
InChi Key: XIRNKXNNONJFQO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OCC
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)