missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Palmitate 97.0+%, TCI America™

Catalog No. P000325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P000325ML 25 mL
P0003500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P000325ML Supplier TCI America Supplier No. P000325ML
Only null left

Chemical Identifiers

CAS 628-97-7
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00008996
InChI Key XIRNKXNNONJFQO-UHFFFAOYSA-N
Synonym ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm
PubChem CID 12366
ChEBI CHEBI:84932
IUPAC Name ethyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC

Specifications

Color White-Yellow
Quantity 25 mL
Formula Weight 284.48
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl Palmitate
EINECSNumber (2)-0798
TSCA Yes