Learn More
Ethyl palmitate, 97%
CAS: 628-97-7 | C18H36O2 | 284.48 g/mol
Supplier: Thermo Scientific Chemicals A1569422
Description
Ethyl palmitate acts as a lubricant on the skinâs surfaces in order to get a soft and smooth appearance. It increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl palmitate acts as a lubricant on the skinâs surfaces in order to get a soft and smooth appearance. It increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol.
Solubility
Miscible with ethanol, dimethyl sulfoxide, chloroform and ethyl acetate. Immiscible with water.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
| Ethyl palmitate | |
| 22°C to 26°C | |
| 192°C to 193°C (10 mmHg) | |
| C18H36O2 | |
| MFCD00008996 | |
| 1782663 | |
| Miscible with ethanol,dimethyl sulfoxide,chloroform and ethyl acetate. Immiscible with water. | |
| CCCCCCCCCCCCCCCC(=O)OCC | |
| 284.48 | |
| CHEBI:84932 | |
| 97% |
| 628-97-7 | |
| 0.857 | |
| 178°C (352°F) | |
| 1.4395 | |
| 100 g | |
| ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm | |
| XIRNKXNNONJFQO-UHFFFAOYSA-N | |
| ethyl hexadecanoate | |
| 12366 | |
| 284.48 |
Chemical Identifiers
| 628-97-7 | |
| 284.48 | |
| XIRNKXNNONJFQO-UHFFFAOYSA-N | |
| 12366 | |
| ethyl hexadecanoate |
| C18H36O2 | |
| MFCD00008996 | |
| ethyl palmitate, hexadecanoic acid, ethyl ester, palmitic acid ethyl ester, ethyl cetylate, palmitic acid, ethyl ester, ethylpalmitate, unii-ird3m534zm, hexadecanoic acid ethyl ester, ethyl n-hexadecanoate, ird3m534zm | |
| CHEBI:84932 | |
| CCCCCCCCCCCCCCCC(=O)OCC |
Safety and Handling
EINECSNumber : 211-064-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only