missing translation for 'onlineSavingsMsg'
Learn More

Ethyl p-Tolylacetate 98.0+%, TCI America™

Catalog No. p-7136667
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12275G 5 g
T122725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12275G Supplier TCI America Supplier No. T12275G

Chemical Identifiers

CAS 14062-19-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00009181
InChI Key BTRGZBIXPLFVNK-UHFFFAOYSA-N
Synonym ethyl p-tolylacetate, ethyl 4-methylphenylacetate, ethyl 2-4-methylphenyl acetate, ethyl 2-p-tolyl acetate, ethyl-p-tolylacetate, p-tolylacetic acid ethyl ester, ethyl-4-tolylacetate, ethyl p-methylphenylacetate, ethyl 4-methylphenyl acetate, 4-methylphenylacetic acid ethyl ester
PubChem CID 84175
IUPAC Name ethyl 2-(4-methylphenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)C

Specifications

Color Yellow
Boiling Point 240°C
Quantity 5 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl p-Tolylacetate