accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 14062-19-2

IUPAC Name: ethyl 2-(4-methylphenyl)acetate
Synonyms: ethyl 2-(4-methylphenyl)acetate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: BTRGZBIXPLFVNK-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results