missing translation for 'onlineSavingsMsg'
Learn More

Ethyl p-Anisate 99.0+%, TCI America™

Catalog No. A093325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A093325ML 25 mL
A0933100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A093325ML Supplier TCI America Supplier No. A093325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94-30-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00015151
InChI Key FHUODBDRWMIBQP-UHFFFAOYSA-N
Synonym ethyl p-anisate, ethyl anisate, ethyl p-methoxybenzoate, p-anisic acid, ethyl ester, benzoic acid, 4-methoxy-, ethyl ester, 4-methoxybenzoic acid ethyl ester, benzoic acid, p-methoxy-, ethyl ester, ethyl p-anisoate, unii-kj95h2s7nm, p-anisic acid ethyl ester
PubChem CID 60979
IUPAC Name ethyl 4-methoxybenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)OC

Specifications

Color Colorless
Boiling Point 143°C
Quantity 25 mL
Formula Weight 180.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl p-Anisate
EINECSNumber (3)-3054
RTECSNumber BZ4697000
TSCA Yes