missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Oxamate 98.0+%, TCI America™

Catalog No. O008525G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O008525G 25 g
O0085250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O008525G Supplier TCI America Supplier No. O008525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 617-36-7
Molecular Formula C4H7NO3
Molecular Weight (g/mol) 117.104
MDL Number MFCD00008005
InChI Key RZMZBHSKPLVQCP-UHFFFAOYSA-N
Synonym ethyl oxamate, ethyl carbamoylformate, oxamic acid ethyl ester, ethyl amino oxo acetate, oxamethane, acetic acid, aminooxo-, ethyl ester, ethoxalamide, oxamidic acid ethyl ester, ethoxy carbonyl formamide, oxalic acid, monoethyl ester amide
PubChem CID 69238
IUPAC Name ethyl 2-amino-2-oxoacetate
SMILES CCOC(=O)C(=O)N

Specifications

Melting Point 114°C
Color White
Quantity 25 g
Formula Weight 117.10
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Oxamate