accessibility menu, dialog, popup

UserName

H3COONH2O

CAS RN 617-36-7

IUPAC Name: ethyl carbamoylformate
Synonyms: ethyl carbamoylformate
Molecular Weight (g/mol): 117.1
Molecular Formula: C4H7NO3
InChi Key: RZMZBHSKPLVQCP-UHFFFAOYSA-N
SMILES: CCOC(=O)C(N)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)