missing translation for 'onlineSavingsMsg'
Learn More

Ethyl O-Mesitylsulfonylacetohydroxamate 98.0+%, TCI America™

Catalog No. M11825G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11825G 5 g
M11821G 1 g
M118225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M11825G Supplier TCI America Supplier No. M11825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38202-27-6
Molecular Formula C13H19NO4S
Molecular Weight (g/mol) 285.358
MDL Number MFCD00009244
InChI Key KQCBSWBQAXTILK-WYMLVPIESA-N
Synonym ethyl o-mesitylsulfonylacetohydroxamate, ethyl n-mesitylsulfonyl oxyacetimidate, ethyl o-methylsulphonylacetohydroxamate, ethyl o-2-mesitylenesulfonyl acethydroxamate, o-mesitylsulfonylacetohydroxamic acid ethyl ester, ethyl 1z-n-mesitylsulfonyl oxy ethanimidoate, ethyl 1e-n-2,4,6-trimethylphenyl sulfonyloxyethanimidate, kqcbswbqaxtilk-owbhpgmisa-n, z-ethyl n-mesitylsulfonyloxyacetimidate, z-ethyl n-mesitylsulfonyl oxyacetimidate
PubChem CID 6282444
IUPAC Name ethyl (1E)-N-(2,4,6-trimethylphenyl)sulfonyloxyethanimidate
SMILES CCOC(=NOS(=O)(=O)C1=C(C=C(C=C1C)C)C)C

Specifications

Melting Point 56°C
Color White
Quantity 5 g
Formula Weight 285.36
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl O-Mesitylsulfonylacetohydroxamate [Precursor of the Powerful Aminating Reagent]
TSCA No
Recommended Storage Refrigerator