missing translation for 'onlineSavingsMsg'
Learn More

Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate, 98+%

Catalog No. p-7036970
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5547006 5 g
AAH5547014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5547006 Supplier Thermo Scientific Chemicals Supplier No. H5547006
Only null left

CAS: 38202-27-6 | C13H19NO4S | 285.358 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38202-27-6
Molecular Formula C13H19NO4S
Molecular Weight (g/mol) 285.358
MDL Number MFCD00009244
InChI Key KQCBSWBQAXTILK-WYMLVPIESA-N
Synonym ethyl o-mesitylsulfonylacetohydroxamate, ethyl n-mesitylsulfonyl oxyacetimidate, ethyl o-methylsulphonylacetohydroxamate, ethyl o-2-mesitylenesulfonyl acethydroxamate, o-mesitylsulfonylacetohydroxamic acid ethyl ester, ethyl 1z-n-mesitylsulfonyl oxy ethanimidoate, ethyl 1e-n-2,4,6-trimethylphenyl sulfonyloxyethanimidate, kqcbswbqaxtilk-owbhpgmisa-n, z-ethyl n-mesitylsulfonyloxyacetimidate, z-ethyl n-mesitylsulfonyl oxyacetimidate
PubChem CID 6282444
IUPAC Name ethyl (1E)-N-(2,4,6-trimethylphenyl)sulfonyloxyethanimidate
SMILES CCOC(=NOS(=O)(=O)C1=C(C=C(C=C1C)C)C)C

Specifications

Melting Point 54°C to 56°C
Quantity 5 g
Beilstein 2144156
Formula Weight 285.36
Percent Purity ≥98%
Chemical Name or Material Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate
EINECSNumber 253-825-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only