missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Nonafluorobutyl Ether (mixture of isomers) 98.0+%, TCI America™

Catalog No. p-7129048
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E052825G 25 g
E0528500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E052825G Supplier TCI America Supplier No. E052825G
Only null left

Chemical Identifiers

CAS 813458-04-7
Molecular Formula C6H5F9O
Molecular Weight (g/mol) 264.091
InChI Key DFUYAWQUODQGFF-UHFFFAOYSA-N
Synonym Ethyl Perfluorobutyl Ether, Nonafluorobutyl Ethyl Ether, Perfluorobutyl Ethyl Ether
PubChem CID 206000
IUPAC Name 1-ethoxy-1,1,2,2,3,3,4,4,4-nonafluorobutane
SMILES CCOC(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Quantity 25 g
Formula Weight 264.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Nonafluorobutyl Ether (mixture of isomers)