accessibility menu, dialog, popup

UserName

CAS RN 813458-04-7

CH3OFFFFFFFFF

CAS RN 813458-04-7

IUPAC Name: 1-ethoxy-1,1,2,2,3,3,4,4,4-nonafluorobutane
Synonyms: ethyl perfluorobutyl ether
Molecular Weight (g/mol): 264.09
Molecular Formula: C6H5F9O
InChi Key: DFUYAWQUODQGFF-UHFFFAOYSA-N
SMILES: CCOC(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results