missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nipecotate, 97%

Catalog No. p-3736495
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC186620250 25 g Glass bottle
AC186621000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC186620250 Supplier Thermo Scientific Chemicals Supplier No. 186620250

CAS: 5006-62-2 | C8H15NO2 | 157.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5006-62-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00005991
InChI Key XIWBSOUNZWSFKU-UHFFFAOYSA-N
Synonym ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate
PubChem CID 98969
IUPAC Name ethyl piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1

Specifications

Density 1.0100g/mL
Boiling Point 102°C to 104°C (7.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4580 to 1.461
Quantity 25 g
Specific Gravity 1.01
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 157.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl nipecotate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 225-681-3

RUO – Research Use Only