missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-(tert-Butoxycarbonyl)-L-pyroglutamate 97.0+%, TCI America™

Catalog No. E11351G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11351G 1 g
E11355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11351G Supplier TCI America Supplier No. E11351G
Only null left

Chemical Identifiers

CAS 144978-12-1
Molecular Formula C12H19NO5
Molecular Weight (g/mol) 257.29
MDL Number MFCD07778090
InChI Key YWWWGFSJHCFVOW-UHFFFAOYNA-N
Synonym boc-pyr-oet, s-ethyl-n-boc-pyroglutamate, n-boc-l-pyroglutamic acid ethyl ester, 1-boc-l-pyroglutamic acid ethyl ester, boc-l-pyroglutamic acid ethyl ester, s-1-tert-butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate, ethyl n-boc-l-pyroglutamate, 1-tert-butyl 2-ethyl 2s-5-oxopyrrolidine-1,2-dicarboxylate, s-n-alpha-t-butyloxycarbonyl-pyroglutamic acid ethyl ester, 1-tert-butyl 2-ethyl s-5-oxopyrrolidine-1,2-dicarboxylate
PubChem CID 5461186
IUPAC Name 1-tert-butyl 2-ethyl 5-oxopyrrolidine-1,2-dicarboxylate
SMILES CCOC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C

Specifications

Melting Point 54°C
Color White
Quantity 1 g
Formula Weight 257.29
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl N-(tert-Butoxycarbonyl)-L-pyroglutamate