missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl N,N-dimethyloxamate, 98%
CAS: 16703-52-9 | C6H11NO3 | 145.158 g/mol
Supplier: Thermo Scientific Chemicals L0892314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl N,N-dimethyloxamate | |
| 1.068 | |
| >110°C (230°F) | |
| 1.443 | |
| 25 g | |
| ethyl n,n-dimethyloxamate, ethyl nn-dimethyloxamate, n,n-dimethyl ethyl oxamate, ethyl 2-dimethylamino-2-oxoacetate, acetic acid, dimethylamino oxo-, ethyl ester, ethyl dimethylcarbamoyl formate, oxamic acid, dimethyl-, ethyl ester, acetic acid, 2-dimethylamino-2-oxo-, ethyl ester, ethyl dimethylamino oxoacetate, aethyl-dimethyloxamat | |
| CCOC(=O)C(=O)N(C)C | |
| 145.158 | |
| 145.16 |
| 16703-52-9 | |
| 118°C to 120°C (10 mmHg) | |
| C6H11NO3 | |
| MFCD00015153 | |
| 1906209 | |
| HMALWDVRMHVUAW-UHFFFAOYSA-N | |
| ethyl 2-(dimethylamino)-2-oxoacetate | |
| 85553 | |
| 98% |
Chemical Identifiers
| 16703-52-9 | |
| 145.158 | |
| HMALWDVRMHVUAW-UHFFFAOYSA-N | |
| 85553 | |
| CCOC(=O)C(=O)N(C)C |
| C6H11NO3 | |
| MFCD00015153 | |
| ethyl n,n-dimethyloxamate, ethyl nn-dimethyloxamate, n,n-dimethyl ethyl oxamate, ethyl 2-dimethylamino-2-oxoacetate, acetic acid, dimethylamino oxo-, ethyl ester, ethyl dimethylcarbamoyl formate, oxamic acid, dimethyl-, ethyl ester, acetic acid, 2-dimethylamino-2-oxo-, ethyl ester, ethyl dimethylamino oxoacetate, aethyl-dimethyloxamat | |
| ethyl 2-(dimethylamino)-2-oxoacetate |
Safety and Handling
EINECSNumber : 240-755-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only