missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N,N-dimethyloxamate, 98%

Catalog No. p-7044971
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0892306 5 g
AAL0892314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0892306 Supplier Thermo Scientific Chemicals Supplier No. L0892306

CAS: 16703-52-9 | C6H11NO3 | 145.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16703-52-9
Molecular Formula C6H11NO3
Molecular Weight (g/mol) 145.158
MDL Number MFCD00015153
InChI Key HMALWDVRMHVUAW-UHFFFAOYSA-N
Synonym ethyl n,n-dimethyloxamate, ethyl nn-dimethyloxamate, n,n-dimethyl ethyl oxamate, ethyl 2-dimethylamino-2-oxoacetate, acetic acid, dimethylamino oxo-, ethyl ester, ethyl dimethylcarbamoyl formate, oxamic acid, dimethyl-, ethyl ester, acetic acid, 2-dimethylamino-2-oxo-, ethyl ester, ethyl dimethylamino oxoacetate, aethyl-dimethyloxamat
PubChem CID 85553
IUPAC Name ethyl 2-(dimethylamino)-2-oxoacetate
SMILES CCOC(=O)C(=O)N(C)C

Specifications

Density 1.068
Boiling Point 118°C to 120°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.443
Quantity 5 g
Beilstein 1906209
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material Ethyl N,N-dimethyloxamate
EINECSNumber 240-755-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only