missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-hydroxyacetimidate, 97%

Catalog No. AAA1100606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1100606 5 g
AAA1100614 25 g
AAA1100622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1100606 Supplier Thermo Scientific Chemicals Supplier No. A1100606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10576-12-2 | C4H9NO2 | 103.12 g/mol

Ethyl N-hydroxyacetimidate was used in the synthesis of O-acyl-and O-nitrophenylhydroxylamines, hydroxamic acid ethoxycarbonylhydrazides and ethyl O-(2,4-dinitrophenyl)acetohydroxamate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl N-hydroxyacetimidate was used in the synthesis of O-acyl-and O-nitrophenylhydroxylamines, hydroxamic acid ethoxycarbonylhydrazides and ethyl O-(2,4-dinitrophenyl)acetohydroxamate.

Solubility
Decompose in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 10576-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00002114,MFCD00002114
InChI Key QWKAVVNRCKPKNM-SNAWJCMRSA-N
Synonym ethyl n-hydroxyacetimidate, n-hydroxyethanimidic acid, ethyl ester, ethyl acetohydroximate, ethyl n-hydroxyethanimidate, e-ethyl n-hydroxyethanimidate, e-ethyl n-hydroxyacetimidate, ethyl 1e-n-hydroxyethanimidate, snx`lfelmmpqh, ethyl acethydroxamate, 1-ethoxyethanone oxime
PubChem CID 6386647
IUPAC Name ethyl (1Z)-N-hydroxyethanimidate
SMILES CCO\C(C)=N\O

Specifications

Melting Point 23°C to 25°C
Boiling Point 55°C to 58°C (6 mmHg)
Flash Point 77°C (171°F)
Refractive Index 1.434
Quantity 5 g
Beilstein 1700260
Sensitivity Moisture sensitive
Solubility Information Decompose in water.
Formula Weight 103.12
Percent Purity 97%
Chemical Name or Material Ethyl N-hydroxyacetimidate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 234-165-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only