missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Myristate 98.0+%, TCI America™

Supplier:  TCI America M0479100ML

 View more versions of this product

Catalog No. M0479100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Myristate
Yellow
C16H32O2
100 mL
MMKRHZKQPFCLLS-UHFFFAOYSA-N
ethyl tetradecanoate
31283
256.43
Liquid
124-06-1
309°C
MFCD00008984
ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
CCCCCCCCCCCCCC(=O)OCC
256.43
CHEBI:84849
≥98.0% (GC)

Chemical Identifiers

124-06-1
256.43
MMKRHZKQPFCLLS-UHFFFAOYSA-N
31283
ethyl tetradecanoate
C16H32O2
MFCD00008984
ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
CHEBI:84849
CCCCCCCCCCCCCC(=O)OCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions