missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Myristate 98.0+%, TCI America™

Catalog No. p-7132193
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M047925ML 25 mL
M0479100ML 100 mL
M0479500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M047925ML Supplier TCI America Supplier No. M047925ML
Only null left

Chemical Identifiers

CAS 124-06-1
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00008984
InChI Key MMKRHZKQPFCLLS-UHFFFAOYSA-N
Synonym ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
PubChem CID 31283
ChEBI CHEBI:84849
IUPAC Name ethyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OCC

Specifications

Color Yellow
Boiling Point 309°C
Quantity 25 mL
Formula Weight 256.43
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Myristate
EINECSNumber (2)-0798
TSCA Yes