missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (Methylthio)acetate 98.0+%, TCI America™
Supplier: TCI America M089725G
Specifications
| Ethyl (Methylthio)acetate | |
| Yellow | |
| C5H10O2S | |
| 25 g | |
| ethyl methylthio acetate, ethyl 2-methylthio acetate, acetic acid, methylthio-, ethyl ester, ethyl 2-methylthio-acetate, ethyl 2-methylsulfanyl acetate, unii-6148qqn25t, ethyl alpha-methylthio acetate, methylthio acetic acid ethyl ester, ethyl .alpha. methylthio acetate, methylsulfanyl-acetic acid ethyl ester | |
| CCOC(=O)CSC | |
| 134.193 | |
| 134.19 | |
| Liquid |
| 4455-13-4 | |
| 176°C | |
| MFCD00009182 | |
| 3272 | |
| MDIAKIHKBBNYHF-UHFFFAOYSA-N | |
| ethyl 2-methylsulfanylacetate | |
| 78199 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 4455-13-4 | |
| 134.193 | |
| MDIAKIHKBBNYHF-UHFFFAOYSA-N | |
| 78199 | |
| CCOC(=O)CSC |
| C5H10O2S | |
| MFCD00009182 | |
| ethyl methylthio acetate, ethyl 2-methylthio acetate, acetic acid, methylthio-, ethyl ester, ethyl 2-methylthio-acetate, ethyl 2-methylsulfanyl acetate, unii-6148qqn25t, ethyl alpha-methylthio acetate, methylthio acetic acid ethyl ester, ethyl .alpha. methylthio acetate, methylsulfanyl-acetic acid ethyl ester | |
| ethyl 2-methylsulfanylacetate |
Safety and Handling
TSCA : No