missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (Methylthio)acetate 98.0+%, TCI America™

Catalog No. M08975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08975G 5 g
M089725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08975G Supplier TCI America Supplier No. M08975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4455-13-4
Molecular Formula C5H10O2S
Molecular Weight (g/mol) 134.193
MDL Number MFCD00009182
InChI Key MDIAKIHKBBNYHF-UHFFFAOYSA-N
Synonym ethyl methylthio acetate, ethyl 2-methylthio acetate, acetic acid, methylthio-, ethyl ester, ethyl 2-methylthio-acetate, ethyl 2-methylsulfanyl acetate, unii-6148qqn25t, ethyl alpha-methylthio acetate, methylthio acetic acid ethyl ester, ethyl .alpha. methylthio acetate, methylsulfanyl-acetic acid ethyl ester
PubChem CID 78199
IUPAC Name ethyl 2-methylsulfanylacetate
SMILES CCOC(=O)CSC

Specifications

Color Yellow
Boiling Point 176°C
Quantity 5 g
UN Number 3272
Formula Weight 134.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (Methylthio)acetate