missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methyl Sulfide 98.0+%, TCI America™

Catalog No. E014325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E014325ML 25 mL
E0143100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E014325ML Supplier TCI America Supplier No. E014325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 624-89-5
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.16
MDL Number MFCD00009268
InChI Key WXEHBUMAEPOYKP-UHFFFAOYSA-N
Synonym ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane
PubChem CID 12230
IUPAC Name (methylsulfanyl)ethane
SMILES CCSC

Specifications

Color Yellow
Boiling Point 66°C
Quantity 25 mL
UN Number 1993
Formula Weight 76.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Methyl Sulfide