missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methyl sulfide, 97%

Catalog No. AAL0285922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0285922 100 g
AAL0285914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0285922 Supplier Thermo Scientific Chemicals Supplier No. L0285922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 624-89-5 | C3H8S | 76.16 g/mol

Ethyl Methyl Sulfide is used as a corrosion inhibitor for zinc metal which finds wide applications like electrode material in batteries, sacrificial anodes and metallic coatings. It is used in mixtures designed to odorize natural gas and is made to by-pass injection system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl Methyl Sulfide is used as a corrosion inhibitor for zinc metal which finds wide applications like electrode material in batteries, sacrificial anodes and metallic coatings. It is used in mixtures designed to odorize natural gas and is made to by-pass injection system.

Solubility
Miscible with alcohols and oils. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 624-89-5
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.16
MDL Number MFCD00009268
InChI Key WXEHBUMAEPOYKP-UHFFFAOYSA-N
Synonym ethyl methyl sulfide, methylthioethane, ethane, methylthio, methyl ethyl sulfide, ethyl methyl sulphide, methyl ethyl sulphide, sulfide, ethyl methyl, unii-mr1gzs62tm, methylthio ethane, methylsulfanyl ethane
PubChem CID 12230
IUPAC Name methylsulfanylethane
SMILES CCSC

Specifications

Melting Point -106°C
Density 0.841
Boiling Point 66°C to 67°C
Flash Point −15°C (5°F)
Odor Stench
Refractive Index 1.4392
Linear Formula CH3CH2SCH3
Quantity 100 g
UN Number UN1993
Beilstein 1696871
Solubility Information Miscible with alcohols and oils. Immiscible with water.
Formula Weight 76.16
Percent Purity 97%
Chemical Name or Material Ethyl methyl sulfide
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 210-868-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only