missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Methoxyacetate 98.0+%, TCI America™
Supplier: TCI America E097925G
Specifications
| Ethyl Methoxyacetate | |
| Yellow | |
| C5H10O3 | |
| 25 g | |
| ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg | |
| CCOC(=O)COC | |
| 118.132 | |
| 118.13 | |
| Liquid |
| 3938-96-3 | |
| 158°C | |
| MFCD00009176 | |
| 3272 | |
| JLEKJZUYWFJPMB-UHFFFAOYSA-N | |
| ethyl 2-methoxyacetate | |
| 77544 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3938-96-3 | |
| 118.132 | |
| JLEKJZUYWFJPMB-UHFFFAOYSA-N | |
| 77544 | |
| CCOC(=O)COC |
| C5H10O3 | |
| MFCD00009176 | |
| ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg | |
| ethyl 2-methoxyacetate |
Safety and Handling
TSCA : No