missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methoxyacetate, 98%

CAS: 3938-96-3 | C5H10O3 | 118.132 g/mol

$333.64 - $333.64

Chemical Identifiers

CAS 3938-96-3
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00009176
InChI Key JLEKJZUYWFJPMB-UHFFFAOYSA-N
Synonym ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg
PubChem CID 77544
IUPAC Name ethyl 2-methoxyacetate
SMILES CCOC(=O)COC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1877018
View Documents
Thermo Scientific Chemicals
A1877018
50 g
Each for $333.64
Only null left
 
Description

Description

Ethyl methoxyacetate was used as acyl donor during the preparation of enantiomers of several phenylethylamines, as acylation reagent for the aminolysis of 1-phenylethanamine and used in an industrial, lipase-catalyzed kinetic resolution of primary amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl methoxyacetate was used as acyl donor during the preparation of enantiomers of several phenylethylamines, as acylation reagent for the aminolysis of 1-phenylethanamine and used in an industrial, lipase-catalyzed kinetic resolution of primary amine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

3938-96-3
118.132
JLEKJZUYWFJPMB-UHFFFAOYSA-N
77544
CCOC(=O)COC
C5H10O3
MFCD00009176
ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg
ethyl 2-methoxyacetate

Specifications

3938-96-3
44°C to 46°C (9 mmHg)
C5H10O3
MFCD00009176
UN3272
ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg
JLEKJZUYWFJPMB-UHFFFAOYSA-N
ethyl 2-methoxyacetate
77544
98%
1
46°C (115°F)
1.401
50 g
1744761
Slightly soluble in water.
CCOC(=O)COC
118.132
118.13
Ethyl methoxyacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 223-521-7

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only