missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methanesulfonate 99.0+%, TCI America™

Catalog No. M060725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M060725G 25 g
M0607100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M060725G Supplier TCI America Supplier No. M060725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62-50-0
Molecular Formula C3H8O3S
Molecular Weight (g/mol) 124.154
MDL Number MFCD00007559
InChI Key PLUBXMRUUVWRLT-UHFFFAOYSA-N
Synonym ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid
PubChem CID 6113
ChEBI CHEBI:23994
IUPAC Name ethyl methanesulfonate
SMILES CCOS(=O)(=O)C

Specifications

Color Yellow
Boiling Point 89°C
Quantity 25 g
Formula Weight 124.15
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Methanesulfonate
RTECSNumber PB2100000
TSCA Yes