missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methanesulfonate, 99%

Catalog No. AAA1293830
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1293830 250 g
AAA1293806 5 g
AAA1293809 10 g
AAA1293818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1293830 Supplier Thermo Scientific Chemicals Supplier No. A1293830

CAS: 62-50-0 | C3H8O3S | 124.154 g/mol

Ethyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.

Solubility
Miscible with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS available online at www.alfa.com.Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 62-50-0
Molecular Formula C3H8O3S
Molecular Weight (g/mol) 124.154
MDL Number MFCD00007559
InChI Key PLUBXMRUUVWRLT-UHFFFAOYSA-N
Synonym ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid
PubChem CID 6113
ChEBI CHEBI:23994
IUPAC Name ethyl methanesulfonate
SMILES CCOS(=O)(=O)C

Specifications

Density 1.17
Boiling Point 85°C to 86°C (10 mmHg)
Flash Point 100°C (212°F)
Odor Fruit-like
Assay Percent Range 99%
Refractive Index 1.418
Quantity 250 g
UN Number UN2810
Beilstein 773969
Sensitivity Moisture sensitive
Merck Index 14,3827
Solubility Information Miscible with water.
Formula Weight 124.16
Percent Purity 99%
Chemical Name or Material Ethyl methanesulfonate
Show More Show Less
Hazard Category H302-H315-H319-H335-H340-H351
Hazard Statement GHS H Statement
H340-H351-H361-H302
May cause genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 200-536-7
RTECSNumber PB2100000
TSCA Yes
storageNote1 Moisture Sensitive
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only