missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl methanesulfonate, 99%
Description
Ethyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.
Solubility
Miscible with water.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS available online at www.alfa.com.Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Ethyl methanesulfonate is used as mutagen for both mammalian and plant cells. It finds application as a model alkylating agent in the study of deoxyribonucleic acid (DNA) repair processes.
Solubility
Miscible with water.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS available online at www.alfa.com.Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Density | 1.17 |
| Boiling Point | 85°C to 86°C (10 mmHg) |
| Flash Point | 100°C (212°F) |
| Odor | Fruit-like |
| Assay Percent Range | 99% |
| Refractive Index | 1.418 |
| Quantity | 5 g |
| UN Number | UN2810 |
| Beilstein | 773969 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319-H335-H340-H351 |
| Hazard Statement | GHS H Statement H340-H351-H361-H302 May cause genetic defects. Suspected of causing cancer. Suspected of damaging fertility or the unborn child. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
| EINECSNumber | 200-536-7 |
| RTECSNumber | PB2100000 |
| TSCA | Yes |
| storageNote1 | Moisture Sensitive |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only