missing translation for 'onlineSavingsMsg'
Learn More

Ethyl maltol, 98%

Catalog No. p-7077351
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450800250 25 g
AC450801000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450800250 Supplier Thermo Scientific Chemicals Supplier No. 450800250
Only null left

CAS: 4940-11-8 | C7H8O3 | 140.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11-8-4940
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.14

Specifications

Melting Point 89.0°C to 93.0°C
Boiling Point 289.0°C to 290.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Merck Index 15,3879
Synonym ethyl maltol, 2-ethyl-3-hydroxy-4h-pyran-4-one, 2-ethyl-3-hydroxy-4-pyrone, veltol plus, 3-hydroxy-2-ethyl-4-pyrone, ethylmaltol, 2-ethylpyromeconic acid, 4h-pyran-4-one, 2-ethyl-3-hydroxy, 3-hydroxy-2-ethyl-1,4-pyrone, 2-ethyl pyromeconic acid
InChI Key YIKYNHJUKRTCJL-UHFFFAOYSA-N
SMILES CCC1=C(C(=O)C=CO1)O
IUPAC Name 2-ethyl-3-hydroxypyran-4-one
PubChem CID 21059
Formula Weight 140.14
Percent Purity 98%
Chemical Name or Material Ethyl maltol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 225-582-5

RUO – Research Use Only