missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Linoleate 97.0+%, TCI America™
$229.10 - $669.97
Chemical Identifiers
| CAS | 544-35-4 |
|---|---|
| Molecular Formula | C20H36O2 |
| Molecular Weight (g/mol) | 308.506 |
| MDL Number | MFCD00009535 |
| InChI Key | FMMOOAYVCKXGMF-MURFETPASA-N |
| Synonym | ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester |
| PubChem CID | 5282184 |
| ChEBI | CHEBI:31572 |
| IUPAC Name | ethyl (9Z,12Z)-octadeca-9,12-dienoate |
| SMILES | CCCCCC=CCC=CCCCCCCCC(=O)OCC |
Chemical Identifiers
| 544-35-4 | |
| 308.506 | |
| FMMOOAYVCKXGMF-MURFETPASA-N | |
| 5282184 | |
| ethyl (9Z,12Z)-octadeca-9,12-dienoate |
| C20H36O2 | |
| MFCD00009535 | |
| ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester | |
| CHEBI:31572 | |
| CCCCCC=CCC=CCCCCCCCC(=O)OCC |
Specifications
| 544-35-4 | |
| 193°C | |
| MFCD00009535 | |
| ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester | |
| CCCCCC=CCC=CCCCCCCCC(=O)OCC | |
| 308.506 | |
| CHEBI:31572 | |
| ≥97.0% (GC) | |
| Ethyl Linoleate |
| Yellow | |
| C20H36O2 | |
| 5 mL | |
| FMMOOAYVCKXGMF-MURFETPASA-N | |
| ethyl (9Z,12Z)-octadeca-9,12-dienoate | |
| 5282184 | |
| 308.51 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0798
RTECSNumber : RG2185000
TSCA : Yes