missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Linoleate 97.0+%, TCI America™

Supplier:  TCI America L013525ML

 View more versions of this product

Catalog No. L013525ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Linoleate
Yellow
C20H36O2
25 mL
FMMOOAYVCKXGMF-MURFETPASA-N
ethyl (9Z,12Z)-octadeca-9,12-dienoate
5282184
308.51
Liquid
544-35-4
193°C
MFCD00009535
ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester
CCCCCC=CCC=CCCCCCCCC(=O)OCC
308.506
CHEBI:31572
≥97.0% (GC)

Chemical Identifiers

544-35-4
308.506
FMMOOAYVCKXGMF-MURFETPASA-N
5282184
ethyl (9Z,12Z)-octadeca-9,12-dienoate
C20H36O2
MFCD00009535
ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester
CHEBI:31572
CCCCCC=CCC=CCCCCCCCC(=O)OCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

RTECSNumber : RG2185000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions