missing translation for 'onlineSavingsMsg'
Learn More

Ethyl levulinate, 98%

CAS: 539-88-8 | C7H12O3 | 144.17 g/mol

$56.33 - $243.62

Chemical Identifiers

CAS 539-88-8
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009209
InChI Key GMEONFUTDYJSNV-UHFFFAOYSA-N
Synonym ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
PubChem CID 10883
IUPAC Name ethyl 4-oxopentanoate
SMILES CCOC(=O)CCC(C)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1500118
View Documents
Thermo Scientific Chemicals
A1500118
50 g
Each for $56.33
Only null left
 
AAA1500122
View Documents
Thermo Scientific Chemicals
A1500122
100 g
Each for $94.62
Only null left
 
AAA1500136
View Documents
Thermo Scientific Chemicals
A1500136
500 g
Each for $243.62
Only null left
 
Description

Description

Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

539-88-8
144.17
GMEONFUTDYJSNV-UHFFFAOYSA-N
10883
CCOC(=O)CCC(C)=O
C7H12O3
MFCD00009209
ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
ethyl 4-oxopentanoate

Specifications

539-88-8
203°C to 205°C
C7H12O3
MFCD00009209
507641
ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
GMEONFUTDYJSNV-UHFFFAOYSA-N
ethyl 4-oxopentanoate
10883
98%
1.014
94°C (201°F)
1.422
50 g
14,3819
Soluble in water.
CCOC(=O)CCC(C)=O
144.17
144.17
Ethyl levulinate
Videos
Safety and Handling

Safety and Handling

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 208-728-2

RTECSNumber : OI1700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only