missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl levulinate, 98%
Description
Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Density | 1.014 |
| Boiling Point | 203°C to 205°C |
| Flash Point | 94°C (201°F) |
| Refractive Index | 1.422 |
| Quantity | 100 g |
| Beilstein | 507641 |
| Merck Index | 14,3819 |
| Solubility Information | Soluble in water. |
| Formula Weight | 144.17 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H227 |
| Precautionary Statement | P210-P280-P370+P378q-P501c |
| EINECSNumber | 208-728-2 |
| RTECSNumber | OI1700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only