missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Laurate 99.0+%, TCI America™
$48.46 - $182.68
Chemical Identifiers
| CAS | 106-33-2 |
|---|---|
| Molecular Formula | C14H28O2 |
| Molecular Weight (g/mol) | 228.38 |
| MDL Number | MFCD00015065 |
| InChI Key | MMXKVMNBHPAILY-UHFFFAOYSA-N |
| Synonym | ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k |
| PubChem CID | 7800 |
| ChEBI | CHEBI:87427 |
| IUPAC Name | ethyl dodecanoate |
| SMILES | CCCCCCCCCCCC(=O)OCC |
Chemical Identifiers
| 106-33-2 | |
| 228.38 | |
| MMXKVMNBHPAILY-UHFFFAOYSA-N | |
| 7800 | |
| ethyl dodecanoate |
| C14H28O2 | |
| MFCD00015065 | |
| ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k | |
| CHEBI:87427 | |
| CCCCCCCCCCCC(=O)OCC |
Specifications
| 106-33-2 | |
| Colorless | |
| C14H28O2 | |
| 25 mL | |
| MMXKVMNBHPAILY-UHFFFAOYSA-N | |
| ethyl dodecanoate | |
| 7800 | |
| 228.38 | |
| Liquid |
| -10°C | |
| 275°C | |
| MFCD00015065 | |
| ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k | |
| CCCCCCCCCCCC(=O)OCC | |
| 228.38 | |
| CHEBI:87427 | |
| ≥99.0% (GC) | |
| Ethyl Laurate |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes