missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Laurate 99.0+%, TCI America™

Catalog No. L001325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L001325ML 25 mL
L0013500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L001325ML Supplier TCI America Supplier No. L001325ML
Only null left

Chemical Identifiers

CAS 106-33-2
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
MDL Number MFCD00015065
InChI Key MMXKVMNBHPAILY-UHFFFAOYSA-N
Synonym ethyl laurate, dodecanoic acid, ethyl ester, ethyl laurinate, ethyl dodecylate, lauric acid ethyl ester, lauric acid, ethyl ester, ethyl laurate natural, dodecanoic acid ethyl ester, ethyl n-dodecanote, unii-f389d4md5k
PubChem CID 7800
ChEBI CHEBI:87427
IUPAC Name ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC

Specifications

Melting Point -10°C
Color Colorless
Boiling Point 275°C
Quantity 25 mL
Formula Weight 228.38
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Laurate
EINECSNumber (2)-0798
TSCA Yes