missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl L-(-)-O-Tosyllactate 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 272.32 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystals |
| Chemical Name or Material | Ethyl L-(-)-O-Tosyllactate |
Safety and Handling
| TSCA | No |