accessibility menu, dialog, popup

UserName

CAS RN 57057-80-4

H3COOH3COSOOCH3

CAS RN 57057-80-4

IUPAC Name: ethyl 2-[(4-methylbenzenesulfonyl)oxy]propanoate
Synonyms: ethyl 2-[(4-methylbenzenesulfonyl)oxy]propanoate
Molecular Weight (g/mol): 272.32
Molecular Formula: C12H16O5S
InChi Key: SNLMUZGICZJWKN-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)OS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results