missing translation for 'onlineSavingsMsg'
Learn More

(-)-Ethyl L-lactate, Analytical Standard, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019810
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019810 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019810 Supplier Merck Emd Millipore Supplier No. 089961ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 687-47-8
Vapor Pressure 5 mmHg (30°C)
Vapor Density 4.1 (vs. air)
Density 1.034 g/mL (at 20°C)
Linear Formula C5H10O3
MDL Number MFCD00004518
Melting Point -26°C
Boiling Point 154°C
Synonym (-)-Ethyl (S)-2-hydroxypropionate; (S)-(-)-2-Hydroxypropionic acid ethyl ester; L-Lactic acid ethyl ester
Quantity 1 mL
Formula Weight 118.13
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less