Learn More
Ethyl isothiocyanate, 96%, Thermo Scientific™
$68.98 - $564.92
Chemical Identifiers
| CAS | 542-85-8 |
|---|---|
| Molecular Formula | C3H5NS |
| Molecular Weight (g/mol) | 87.14 |
| MDL Number | MFCD00004820 |
| InChI Key | HBNYJWAFDZLWRS-UHFFFAOYSA-N |
| Synonym | ethyl isothiocyanate, ethane, isothiocyanato, ethyl mustard oil, ethylisothiocyanate, isothiocyanic acid, ethyl ester, unii-3284mj2t8p, ccris 7323, ethylisothio-cyanate, isothiocyanato-ethane |
| PubChem CID | 10966 |
| ChEBI | CHEBI:85098 |
| IUPAC Name | isothiocyanatoethane |
| SMILES | CCN=C=S |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC376860050
|
N/A | 5g | Glass bottle |
Each for $68.98
|
|
||||
|
AC376860250
|
N/A | 25g | Glass bottle |
Each for $216.05
|
|
||||
|
AC376861000
|
N/A | 100g | Glass bottle |
Each for $564.92
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C3H5NS | |
| MFCD00004820 | |
| ethyl isothiocyanate, ethane, isothiocyanato, ethyl mustard oil, ethylisothiocyanate, isothiocyanic acid, ethyl ester, unii-3284mj2t8p, ccris 7323, ethylisothio-cyanate, isothiocyanato-ethane | |
| CHEBI:85098 | |
| CCN=C=S |
Specifications
| 542-85-8 | |
| 130°C to 132°C | |
| Authentic | |
| C3H5NS | |
| C2H5NCS | |
| 0.995 | |
| ethyl isothiocyanate, ethane, isothiocyanato, ethyl mustard oil, ethylisothiocyanate, isothiocyanic acid, ethyl ester, unii-3284mj2t8p, ccris 7323, ethylisothio-cyanate, isothiocyanato-ethane | |
| HBNYJWAFDZLWRS-UHFFFAOYSA-N | |
| isothiocyanatoethane | |
| 10966 | |
| 87.14 | |
| Ethyl isothiocyanate |
| 0.9950g/mL | |
| 32°C | |
| 95% min. (GC) | |
| 1.5100 to 1.513 | |
| MFCD00004820 | |
| 15, 3870 | |
| Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether | |
| CCN=C=S | |
| 87.14 | |
| CHEBI:85098 | |
| 96% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Causes serious eye damage.
Toxic in contact with skin.
Flammable liquid and vapor.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 208-831-2
RUO – Research Use Only