missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isopropyl Sulfide 98.0+%, TCI America™

Catalog No. p-7128898
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02255G 5 g
E022525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E02255G Supplier TCI America Supplier No. E02255G

Chemical Identifiers

CAS 5145-99-3
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.211
MDL Number MFCD00039908
InChI Key NZUQQADVSXWVNW-UHFFFAOYSA-N
PubChem CID 21228
IUPAC Name 2-ethylsulfanylpropane
SMILES CCSC(C)C

Specifications

Color Colorless
Boiling Point 103°C
Quantity 5 g
UN Number 1993
Formula Weight 104.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Isopropyl Sulfide