missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isopropyl Ether 98.0+%, TCI America™

Catalog No. E041610ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E041610ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E041610ML Supplier TCI America Supplier No. E041610ML
Only null left

Chemical Identifiers

CAS 625-54-7
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00043532
InChI Key XSJVWZAETSBXKU-UHFFFAOYSA-N
Synonym 2-Ethoxypropane
PubChem CID 12256
IUPAC Name 2-ethoxypropane
SMILES CCOC(C)C

Specifications

Color Colorless
Boiling Point 51°C
Quantity 10 mL
UN Number 3271
Formula Weight 88.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Isopropyl Ether
TSCA Yes
Recommended Storage Refrigerator