accessibility menu, dialog, popup

UserName

CH3OCH3H3C

CAS RN 625-54-7

IUPAC Name: 2-ethoxypropane
Synonyms: ethyl isopropyl ether
Molecular Weight (g/mol): 88.15
Molecular Formula: C5H12O
InChi Key: XSJVWZAETSBXKU-UHFFFAOYSA-N
SMILES: CCOC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results