missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isocyanoacetate 97.0+%, TCI America™

Supplier:  TCI America I056225G

 View more versions of this product

Catalog No. I056225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethyl Isocyanoacetate
Yellow
C5H7NO2
25 g
ethyl isocyanoacetate, isocyanoacetic acid ethyl ester, ethylisocyanoacetate, acetic acid, 2-isocyano-, ethyl ester, acetic acid, isocyano-, ethyl ester, ethyl iso-cyanoacetate, ethyl isocyano-acetate, pubchem2345, ethyl 2-isocyanoacetat
CCOC(=O)C[N+]#[C-]
113.12
113.12
Liquid
2999-46-4
90°C
MFCD00000007
2810
FPULFENIJDPZBX-UHFFFAOYSA-N
ethyl 2-isocyanoacetate
533707
≥97.0% (GC)

Chemical Identifiers

2999-46-4
113.12
FPULFENIJDPZBX-UHFFFAOYSA-N
533707
CCOC(=O)C[N+]#[C-]
C5H7NO2
MFCD00000007
ethyl isocyanoacetate, isocyanoacetic acid ethyl ester, ethylisocyanoacetate, acetic acid, 2-isocyano-, ethyl ester, acetic acid, isocyano-, ethyl ester, ethyl iso-cyanoacetate, ethyl isocyano-acetate, pubchem2345, ethyl 2-isocyanoacetat
ethyl 2-isocyanoacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4114

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions