missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isocyanoacetate 97.0+%, TCI America™

Catalog No. p-7131485
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05625G 5 g
I056225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05625G Supplier TCI America Supplier No. I05625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2999-46-4
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00000007
InChI Key FPULFENIJDPZBX-UHFFFAOYSA-N
Synonym ethyl isocyanoacetate, isocyanoacetic acid ethyl ester, ethylisocyanoacetate, acetic acid, 2-isocyano-, ethyl ester, acetic acid, isocyano-, ethyl ester, ethyl iso-cyanoacetate, ethyl isocyano-acetate, pubchem2345, ethyl 2-isocyanoacetat
PubChem CID 533707
IUPAC Name ethyl 2-isocyanoacetate
SMILES CCOC(=O)C[N+]#[C-]

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
UN Number 2810
Formula Weight 113.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Isocyanoacetate
EINECSNumber (2)-4114
TSCA No
Recommended Storage Refrigerator