missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isocyanoacetate, 98%

CAS: 2999-46-4 | C5H7NO2 | 113.12 g/mol

Supplier:  Thermo Scientific Chemicals L1537906

 View more versions of this product

Catalog No. AAL1537906


Only null left
Explore available promotions

Description

Description

Ethyl isocyanoacetate is used in the syntheses of 7-aza-tetrahydroindoles, oxazolines, benzodiazepines, imidazoles and oxazoles. For example, it is involved in the synthesis of 1H-indole-2-carboxylate by condensing with 2-bromo benzenealdehyde using CuI as a catalyst. It is a precursor used in the preparation of ethyl thiazole-4-carboxylate upon reaction with O-ethyl thioformate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl isocyanoacetate is used in the syntheses of 7-aza-tetrahydroindoles, oxazolines, benzodiazepines, imidazoles and oxazoles. For example, it is involved in the synthesis of 1H-indole-2-carboxylate by condensing with 2-bromo benzenealdehyde using CuI as a catalyst. It is a precursor used in the preparation of ethyl thiazole-4-carboxylate upon reaction with O-ethyl thioformate.

Solubility
Miscible with organic solvents. Slightly miscible with water.

Notes
Store in cool place. Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Ethyl isocyanoacetate
1.035
84°C (183°F)
1.422
5 g
3588613
ethyl isocyanoacetate, isocyanoacetic acid ethyl ester, ethylisocyanoacetate, acetic acid, 2-isocyano-, ethyl ester, acetic acid, isocyano-, ethyl ester, ethyl iso-cyanoacetate, ethyl isocyano-acetate, pubchem2345, ethyl 2-isocyanoacetat
FPULFENIJDPZBX-UHFFFAOYSA-N
ethyl 2-isocyanoacetate
533707
98%
2999-46-4
194°C to 196°C
C5H7NO2
MFCD00000007
UN2810
Light and moisture sensitive
Miscible with organic solvents. Slightly miscible with water.
CCOC(=O)C[N+]#[C-]
113.12
113.12

Chemical Identifiers

2999-46-4
113.12
FPULFENIJDPZBX-UHFFFAOYSA-N
533707
CCOC(=O)C[N+]#[C-]
C5H7NO2
MFCD00000007
ethyl isocyanoacetate, isocyanoacetic acid ethyl ester, ethylisocyanoacetate, acetic acid, 2-isocyano-, ethyl ester, acetic acid, isocyano-, ethyl ester, ethyl iso-cyanoacetate, ethyl isocyano-acetate, pubchem2345, ethyl 2-isocyanoacetat
ethyl 2-isocyanoacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 221-077-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only