missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isobutyrate, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193447
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019888 1 mL
111018400 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019888 Supplier Merck Emd Millipore Supplier No. 195361ML
Only null left

Specifications

CAS 97-62-1
Vapor Pressure 40 mmHg (33.8°C)
Vapor Density 4.01 (vs. air)
Density 0.865 g/mL (at 25°C)
Physical Form Neat
Linear Formula (CH3)2 CHCOOC2H5
MDL Number MFCD00009165
Boiling Point 112°C to 113°C
Quantity 1 mL
Formula Weight 116.16
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less